missing translation for 'onlineSavingsMsg'
Learn More

Manutan Vinyl tape, white, width 12mm

Brand:  Manutan A329008

Encompass_Preferred

Product Code. 12807249

  • 23.23 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use to seal and colour code cases and cartons, making identification during storage or transit easy. Good adhesion, resistant to tearing and ripping. Glossy top surface, non-stretch and difficult to tear or burst. Perform well in deep freeze applications.
  • Length : 66m
  • Width : 12mm
  • Pack quantity : 12
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers