missing translation for 'onlineSavingsMsg'
Learn More
Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10759344

Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10759344
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10759344

Brand: Thermo Scientific Acros 209391000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-67-1, 109-99-9
Molecular Formula C2H3BrMg
Molecular Weight (g/mol) 131.26
MDL Number MFCD00000042
InChI Key XHHHAXOHMKAOSL-UHFFFAOYSA-M
Synonym vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide, grignard reagent
PubChem CID 74584
SMILES Br[Mg]C=C

Specifications

Color Brown
Linear Formula H2C=CHMgBr
Concentration 0.7 to 0.9M
Density 0.9800g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name bromo(ethenyl)magnesium
Formula Weight 131.25
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.98
Quantity 100 mL
Flash Point −17°C
Physical Form Solution
Chemical Name or Material Vinylmagnesium bromide, 0.7M Solution in THF
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468375 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number209398000,209391000
Lot NumberA0457167 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number209398000,209391000
Lot NumberA0457167 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog Number209398000,209391000
Lot NumberA0451379 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number209398000,209391000
Lot NumberA0447934 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number209398000,209391000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylmagnesium bromide, 0.7M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.