missing translation for 'onlineSavingsMsg'
Learn More

Vinylmagnesium chloride, 1.7M (15 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

81.00 € - 442.00 €

Chemical Identifiers

CAS 3536-96-7, 109-99-9
Molecular Formula C2H3ClMg
Molecular Weight (g/mol) 86.80
MDL Number MFCD00013594
InChI Key NAFZXKWEUWVSAW-UHFFFAOYSA-M
Synonym vinylmagnesium chloride, chlorovinylmagnesium, chloro ethenyl magnesium, vinyl magnesium chloride, magnesium, chloroethenyl, vinyl magnesium-chloride, vinyl-magnesium chloride, magnesium,chloroethenyl, ethenyl magnesium chloride, vinylmagnesium chloride solution
PubChem CID 77075
IUPAC Name magnesium;ethene;chloride
SMILES Cl[Mg]C=C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10507763
View Documents
Thermo Scientific Acros
252591000
100 mL AcroSeal™ Glass bottle
81.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10043260
View Documents
Thermo Scientific Acros
252598000
800 mL AcroSeal™ Glass Bottle 442.00 €
800mL
Qty Price Break Available
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3536-96-7, 109-99-9
86.80
NAFZXKWEUWVSAW-UHFFFAOYSA-M
77075
Cl[Mg]C=C
C2H3ClMg
MFCD00013594
vinylmagnesium chloride, chlorovinylmagnesium, chloro ethenyl magnesium, vinyl magnesium chloride, magnesium, chloroethenyl, vinyl magnesium-chloride, vinyl-magnesium chloride, magnesium,chloroethenyl, ethenyl magnesium chloride, vinylmagnesium chloride solution
magnesium;ethene;chloride

Specifications

White to Beige
66.0°C
CH2=CHMgCl
0.9800g/mL
Solubility in water: vigorous reaction.
Cl[Mg]C=C
86.80
86.8
0.98
Crystalline Powder
3536-96-7, 109-99-9
C2H3ClMg
MFCD00013594
vinylmagnesium chloride, chlorovinylmagnesium, chloro ethenyl magnesium, vinyl magnesium chloride, magnesium, chloroethenyl, vinyl magnesium-chloride, vinyl-magnesium chloride, magnesium,chloroethenyl, ethenyl magnesium chloride, vinylmagnesium chloride solution
NAFZXKWEUWVSAW-UHFFFAOYSA-M
magnesium;ethene;chloride
77075
2M (18 wt.%)
−017°C
Vinylmagnesium chloride, 15 wt. % Solution in Tetrahydrofuran
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

missing translation for 'einecsNumber' : 222-575-9

Recommended Storage : Below 10°C, crystallization of Grignard reagent will occur. Store between 10°C and 25°C.

Product Identifier
  • Vinylmagnesium chloride, 1.9M (16.5 wt.%) solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.