Learn More
Vinyltrimethylsilane, 98+%, Thermo Scientific Chemicals
40.30 € - 416.00 €
Chemical Identifiers
CAS | 754-05-2 |
---|---|
Molecular Formula | C5H12Si |
Molecular Weight (g/mol) | 100.236 |
MDL Number | MFCD00008606 |
InChI Key | GCSJLQSCSDMKTP-UHFFFAOYSA-N |
Synonym | vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane |
PubChem CID | 79102 |
IUPAC Name | ethenyl(trimethyl)silane |
SMILES | C[Si](C)(C)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343886
|
Thermo Scientific Alfa Aesar
L16989.06 |
5 g |
40.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353886
|
Thermo Scientific Alfa Aesar
L16989.14 |
25 g |
134.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363886
|
Thermo Scientific Alfa Aesar
L16989.22 |
100 g |
416.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
754-05-2 | |
100.236 | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
79102 | |
C[Si](C)(C)C=C |
C5H12Si | |
MFCD00008606 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
ethenyl(trimethyl)silane |
Specifications
54°C to 56°C | |
754-05-2 | |
MFCD00008606 | |
956572 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
C[Si](C)(C)C=C | |
100.236 | |
100.24 | |
Mild |
-132°C | |
C5H12Si | |
UN1993 | |
−24°C (−11°F) | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
ethenyl(trimethyl)silane | |
79102 | |
≥98% | |
Vinyltrimethylsilane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P280g-P243-P305+P351+P338-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep container tightly closed.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only