missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Viral Production Cells 2.0
Click to view available options
Quantity:
1 mL
6 x 1 mL
Unit Size:
Each
Pack of 6
Description
Gibco Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System.
- Adapted to the chemically defined Viral Production Medium and maintained in suspension culture in that medium
- Highly transfectable using the AAV-MAX Transfection Kit for adeno-associated virus (AAV) production
- Supplied frozen in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Viral Production Medium and 10% DMSO
- Should be thawed directly into Viral Production Medium supplemented with 4 mM GlutaMAX Supplement (Cat. No. 35050038) for optimal performance
- Clonal, 293F-derived, high-producer cell line
- Absence of SV40 large T antigen
- Optimized for high-density, suspension culture (>12 million cells/mL) in chemically defined medium
- Enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
- Robust scalability and passage stability
Specifications
Specifications
| Cell State | Frozen |
| Concentration | 1e7 cells/mL |
| Content And Storage | 6 cryovials, store in vapor phase liquid nitrogen |
| Cell Line | Viral Production Cells 2.0 |
| Cell Type | Kidney (Embryonic) |
| Culture Type | Suspension Cell Culture |
| Form | Frozen |
| Species | Human |
| Format | 6 Vials |
| For Use With (Application) | AAV Production |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction