Learn More
Applied Biosystems™ ViralSEQ™ Vesivirus Detection Kit
The ViralSEQ Vesivirus Detection Assay is a PCR-based test that enables users to quickly isolate and detect Vesivirus 2117 and all other known strains with high sensitivity and specificity.
Brand:  Applied Biosystems™ 4448398C
Product Code. 30000685
  • 3235.00 € / 100 reactions
Explore more special offers
Description
Show More
Specifications
Specifications
Contains RT-PCR assay, buffer and enzyme mix, positive control and negative control
RNA
100 Reactions
Vesivirus Detection Kit
Dry Ice
Primer-probe
1-Step qRT-PCR
ViralSEQ™
100 reactions
Viral RNA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2407009 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number4448398C
Lot Number2401008 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number4448398C
Lot Number2304007 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number4448398C
Lot Number2205005 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog Number4448398C
Lot Number2109004 Certificate TypeCertificate of Analysis Date22/09/2021 Catalog Number4448398C
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ ViralSEQ™ Vesivirus Detection Kit > 100 Reactions