missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Day Impex™ Virkon™ Virucidal Disinfectant
Clean and disinfect work surfaces and equipment in a single, one-step operation. Day Impex™ Virkon™ Virucidal Disinfectant provides the widest proven spectrum of any disinfectant.
708.00 € - 826.80 €
Specifications
Product Type | Virucidal Disinfectant |
---|---|
For Use With (Application) | Virucidal Disinfectant |
Alert:
Use biocides safely. Always read the label and product information before use
Product Code | Brand | Volume (Metric) | Container Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Container Type | Quantity | Price | Quantity & Availability | |||
12338667
|
Day Impex™
330001 |
50 g | Sachet | 50/Pk. |
744.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12348667
|
Day Impex™
330002 |
500 g | Squeeze Pack | 6/Pk. |
708.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12358667
|
Day Impex™
330003 |
5 kg | Drum | 1 Ea. |
826.80 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Use biocides safely. Always read the label and product information before use
- Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
- Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
- Can be applied in powder form direct to spillages of blood and other body fluids
- Easily dissolves to make a liquid disinfectant
- Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
- Under COSHH requirements, no maximum exposure limit is required
- Non-corrosive, non-bleaching and biodegradable
- Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application
- For cleaning and disinfecting work surfaces and equipment
Specifications
Virucidal Disinfectant | |
Virucidal Disinfectant |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : pentapotassium bis(peroxymonosulphate)bis(sulphate), benzenesulphonic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Day Impex™ Virkon™ Virucidal Disinfectant