Learn More
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Designed specifically to offer outstanding comfort, flexibility and good chemical resistance. Ansell Virtex™ 79-700 Series Blue Nitrile Immersion Gloves provide all-day comfort in a lightweight glove.
Brand:  Ansell™ 79700080
 View more versions of this product
Product Code. 11736703
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Reusable
  • Anti-static according to EN1149
  • With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
  • Significantly lower tendency to shed than traditional cotton flock gloves
  • Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrile
Immersion Gloves
79-700
Blue
Aquadri
Good chemical resistance
Nitrile
8
0.2 mm
Straight
50 Pairs
79-700