Learn More
3M™ Virtua™ AP Safety Glasses
Click to view available options
Frame Color: Clear; Lens Tint: Clear
Description
- Sleek, unisex-styled safety glasses
- Comfortable, impact-resistant, lightweight frame
- High-wrap lens fits snug against the face for better coverage
- Integral side shields and browguard
- The modern design offers excellent coverage and field of vision
Specifications
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Spectacles |
No. per Pack | 20 pairs |