Learn More
SI Analytics™ ViscoPump III
SI Analytics™ ViscoPump III comes equipped with a connection cable.
Brand:  SI Analytics™ 285424060
 View more versions of this product
Product Code. 15705078
  • 4805.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optical Detection: Connection cable for measuring stand, tube holder VZ 5127 and holder for safety bottle
  • Thermal Detection: Connection cable for TC viscometers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ViscoPump III
Optical Detection