missing translation for 'onlineSavingsMsg'
Learn More

Viscosity Standard, 11mm2/sec., Poulten Selfe™

Product Code. 13131123
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
  • PSL has shown that it meets ISO 17025 and has been accredited to manufacture, calibrate and certify viscosity oils by UKAS. Kinematic viscosity oil standards as specified by ISO 3104-3105, ASTM D445-D446, IP71, ASTM D2170, BS 188.Nominal viscosities (mm2s-1, cSt) - kinematic. Nominal viscosities (mPas, cP) - dynamic.
  • Viscosity oil standards
    PSL viscosity oil standards are supplied with UKAS certificates of calibration providing the user with official assurance of the competence of the calibration work carried out. UKAS certificates of calibration provide several temperatures as shown on the table below together with uncertainties of measurement. PSL viscosity oil standards, supplied in 500mL containers, are Newtonian in character and are suitable for use with many types of viscometers including, in particular, glass capillary and rotational viscometers as well as flow cups.

Specifications

Quantity 500 mL
Viscosity 11mm2/sec. at 20°C Kinematic Viscosity
compliance-icons
Product Identifier
  • 500 ML Viscosity oil standards kinematic / dynamic
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C11-C14
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'