missing translation for 'onlineSavingsMsg'
Learn More

Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100, Thermo Scientific Chemicals

Product Code. 15447365
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 8042-47-5
Quantity 500 mL
Molecular Formula C16H10N2Na2O7S2
MDL Number MFCD00084411
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 452.363
PubChem CID 9566064
Grade Specpure™
Odor Odorless
Flash Point >110°C (230°F)
Physical Form Liquid
Chemical Name or Material Viscosity standard, Nominally 50cSt at 40 and 7.3cSt at 100
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'