missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Viscous Sampler

Operates like a large syringe by having the operator pull the handle up to suck the product into the sampler. Sampling Systems™ Viscous Sampler is ideal for sampling creams, shampoos, soaps, honey and other medium viscous materials.

Brand:  Sampling Systems™ 7000P-2000

 View more versions of this product

Product Code. 15829381

  • 635.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for medium viscosity materials.
  • Manufactured from PP, 316L Stainless Steel or PTFE
  • Works like a large a syringe
  • The operator pulls up the handle in order to suck the product into the sampler
Specifications

Specifications

Viscous Fluid Sampler
650 mL
25 mm
Polypropylene
Medium Viscosity Materials
2000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers