Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Polarizer Set Only for Ring Light S40-75
The VisiLED Polarizer Set, with cutting edge polarization filters, is specially designed for the S40-75.
Brand:  SCHOTT AG Lighting and Imaging™ 400575
Product Code. 16677105
  • 605.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polarization system for VisiLED Brightfield Ring Light S40-75
  • Turnable polarizer with adapting thread and sliding filter as analyzer
  • Polarized light by an polarizing and analyzing filter within the beam path can be used very efficiently used in Stereo Microscopy to contrast inner structure of material or to identify metal particles within others
  • Not for use with any other ringllight
  • To use only with VisiLED Ring Light S40-75
Specifications
Specifications
Polarization Filter
A Wide Variety of Tasks From Life Science to Industrial Applications
Polarizer Filter and Analyzer
Polarizer M90 x 1 Analyzer for 66 mm lens
VisiLED Ringlight S40-75
M90 x 1