missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Pipette Tips: Low Retention
For use with Ovation and MLA Brand pipetters
Brand: VistaLab™ Technologies 4070-1032LR
This item is not returnable.
View return policy
Description
- Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
- Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Low Retention | |
Non-filtered | |
Clear | |
Autoclavable | |
5 in. | |
e-beam | |
Low Retention, Straight | |
192 Tips/Rack | |
Ovation | |
Polypropylene |
25 μL | |
Sterile | |
Blue | |
1.7 in. | |
Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications | |
Rack | |
VistaRak: 5 racks | |
Single-use | |
No |