missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoirs
Unique trough-within-trough design maximizes reagent recovery
Brand: VistaLab™ Technologies 4054-1002
This item is not returnable.
View return policy
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
- 100mL model (Cat. No. 21-385-111) is supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Order Info
Reservoir lid sold separately (Cat. No. 21-385-110).
Specifications
Pyrogen-free | |
25 mL | |
Non-sterile | |
7.5 in. | |
Bulk | |
7 in. |
White | |
Polystyrene | |
Reagent Reservoir | |
13 in. | |
10/bag x 10; 100/Cs. | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction