Learn More
Cole Parmer Vistek™ Vibration Isolation Platforms
Significantly improve the performance of microscopes, lab balances, and other sensitive instruments
Brand:  Cole Parmer 11500-03
2422.96 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 11858651
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Cut off the transmission of vibrations from the tabletop or countertop. Effectively isolate horizontal and vertical vibration components in all six degrees of freedom.
  • Passive, mechanical and maintenance-free No air hoses or supporting hardware required
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole Parmer Vistek™ Vibration Isolation Platforms > For lab Balances; Steel; Load Range: 0 to 44 lbs (0 to 20kg)