Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Acidic
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
Brand: R&D Systems™ VCTS022
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Unlike Universal solution, Acidic and Basic solutions may dramatically enhance immunoreactivity but may also affect tissue morphology.
- The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
Spezifikation
Store the unopened product at room temperature. Do not use past expiration date. | |
50 mL |