Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Universal
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
Brand:  R&D Systems™ VCTS023
Product Code. 30002154
  • 150.00 € / 50mL
Explore more special offers
Description
Description
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store the unopened product at room temperature. Do not use past expiration date.
50 mL