missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin A acetate in gelatin, 500,000 I.U./g, Thermo Scientific Chemicals
32.60 € - 325.00 €
Chemical Identifiers
CAS | 127-47-9 |
---|---|
Molecular Formula | C22H32O2 |
Molecular Weight (g/mol) | 328.496 |
MDL Number | MFCD00019413 |
InChI Key | QGNJRVVDBSJHIZ-QHLGVNSISA-N |
Synonym | retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 |
PubChem CID | 638034 |
ChEBI | CHEBI:32095 |
IUPAC Name | [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate |
SMILES | CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453347
|
Thermo Scientific Alfa Aesar
A16237.14 |
25 g |
32.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463347
|
Thermo Scientific Alfa Aesar
A16237.22 |
100 g |
84.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473347
|
Thermo Scientific Alfa Aesar
A16237.36 |
500 g |
325.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-47-9 | |
328.496 | |
QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
638034 | |
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate |
C22H32O2 | |
MFCD00019413 | |
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 | |
CHEBI:32095 | |
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C |
Specifications
127-47-9 | |
MFCD00019413 | |
Air and light sensitive; Hygroscopic | |
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650 | |
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C | |
328.496 | |
CHEBI:32095 | |
Vitamin A acetate, in gelatin |
C22H32O2 | |
1915439 | |
14,10013 | |
QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate | |
638034 | |
500,000 IU/g |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360D
missing translation for 'einecsNumber' : 204-844-2
missing translation for 'rtecsNumber' : VH6825000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only