missing translation for 'onlineSavingsMsg'
Learn More
Vitamin E acetate, 97%, Thermo Scientific Chemicals Product Code.: 11461848

Vitamin E acetate, 97%, Thermo Scientific Chemicals

Product Code. 11461848
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11461848

Brand: Thermo Scientific Alfa Aesar A14505.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

Solubility
Immiscible with water.

Notes
Store in a cool place. Air and light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.754
MDL Number MFCD00072042
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point °C to 28°C
Density 0.960
Boiling Point >300°C
Flash Point 210°C (410°F)
Assay Percent Range 97%
Refractive Index 1.496
Quantity 25 g
Beilstein 97512
Sensitivity Air and light sensitive
Merck Index 14,9495
Solubility Information Immiscible with water.
Formula Weight 472.76
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vitamin E Acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP03L021 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog NumberA14505.14,A14505.36,A14505.22
Lot NumberY18K036 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog NumberA14505.14,A14505.36,A14505.22
Lot NumberS20K044 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog NumberA14505.14,A14505.36,A14505.22
Lot NumberR30K033 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog NumberA14505.14,A14505.36,A14505.22
Lot NumberM04J010 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog NumberA14505.14,A14505.36,A14505.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vitamin E acetate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.