Learn More
Fisherbrand™ Viton Rubber Tubing
Suitable for transferring corrosive liquids, even at high temperatures
Brand:  Fisherbrand™ FB68946
 View more versions of this product
Product Code. 11572583
  • 143.00 € / 5m
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Excellent chemical resistance, particularly to aggressive fuels, acids, animal and vegetable oils, solvents, alcohols and bleaching agents
  • Good resistance to ozone and radiation
  • Other sizes are available on request
Specifications
Specifications
6 mm
5 m
Transferring corrosive liquids, even at high temperatures
Viton
1.5 mm