Learn More
Heidolph instruments Viton™ Tubing
Use this tubing when you require excellent acid resistance at high temperatures. Heidolph™ Viton™ Tubing is smooth, biocompatible and inert.
Brand:  Heidolph instruments 525-53000-00
 View more versions of this product
Product Code. 10065751
  • 73.00 € / 1m
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low gas permeability.
  • Resistant to solvents and corrosives at high temperatures.
Specifications
Specifications
Autoclavable
1.6 mm
0.8
Borosilicate Glass
Non-sterile
0.7/1/7 bar