missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Vitronectin (VTN-N) Recombinant Human Protein, Truncated

Product Code. p-7182898
Click to view available options
Quantity:
1 mL = 0.5 mg
10 mL = 5 mg
Unit Size:
1 unit
1mL
This item is not returnable. View return policy

Product Code. 15134499

Brand: Gibco™ A14700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs).

Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8 Medium (Cat. No. A1517001), VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.

  • Proven to maintain pluripotency and normal growth characteristics in multiple PSC lines when used with Essential 8 Medium
  • Supports PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages
  • Supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8 Medium
  • Optimized—designed in the laboratory of James Thomson for use with Essential 8 Medium
  • Defined—for reduced variability in your PSC cultures
  • Supports human pluripotent stem cell attachment and survival better than wild type vitronectin
  • Optimal environment for feeder-free pluripotent stem cell culture
  • Defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs)
TRUSTED_SUSTAINABILITY

Specifications

Concentration 500 μg/mL
For Use With (Application) Stem Cell Culture
Purification Method HPLC
Quantity 1 mL = 0.5 mg
Species Human
Recombinant Recombinant
Content And Storage Store at -80°C and protect from light.
Classification AOF
Endotoxin Level Very Low
Shipping Condition Dry ice
Volume (Metric) 1 mL
Protein Form Recombinant
Form Liquid
Protein Subtype Vitronectin
Research Category Stem Cell Research
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.