missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint
Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.
Brand: Vitlab™ 1332819
This item is not returnable.
View return policy
Description
- Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
- VENT-CAP practically eliminates leakage and prevents almost all static overpressure
- Color-coded for easy visual identification of the media
- Material name in German, English, French and Spanish
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications
500 mL | |
Saving and storing | |
LDPE | |
Red | |
Narrow | |
Wash Bottle | |
74 mm |
Imprint: Distilled Water | |
Spray tube and imprint | |
12 Pack | |
Yes | |
180 mm | |
Polypropylene | |
Vented |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction