missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Vivacon 500 125kDa
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Vivacon 500 125kDa are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples.
Brand: Sartorius Stedim Biotech™ VN01H82
476.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The product of choice for protein fractionation applications.
- The use of Hydrosart™ membranes results in high recoveries from dilute samples.
- Complete and highly reproducible recovery of the concentrate.
- Reverse spinning step capacity.
- Concentration of up to 0.5 mL genomic DNA or peptide samples.
- Low binding material.
- MWCO printed on device.
- Selected cut offs available sterile and DNA-free for things like forensic applications.
- qPCR assay over 55 cycles with a verified limit of detection <2 pg
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
0.1 to 0.5 mL, 5 μL (Dead Stop Volume) | |
Centrifugal ultrafiltration device | |
Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane | |
Single-use |
100 Pk. | |
Sterile | |
Yes | |
Centrifugal Units |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction