Learn More
Sartorius Vivaspin™ 20 Centrifugal Concentrator
Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.
225.00 € - 882.00 €
Specifications
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
Material (Membrane) | PES |
Sample Volume | 20 mL |
Sterility | Non-sterile |
Product Code | Brand | MWCO | Quantity | Pore Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Pore Size | Price | Quantity & Availability | ||||||
10757651
![]() |
Sartorius
VS2042 |
100 kDa | 48/Pk. |
825.00 €
Pack of 48 |
In Stock
Log in to see stock. ![]() |
|||||||
10365171
|
Sartorius
VS2062 |
1000 kDa | 48/Pk. | N/A | N/A | N/A | ||||||
10506282
![]() |
Sartorius
VS2021 |
30 kDa | 12/Pk. |
246.00 €
Pack of 12 |
In Stock
Log in to see stock. ![]() |
|||||||
10623002
|
Sartorius
VS20S1 |
Starter Pack 2 each of: 5K, 10K, 30K, 50K, 100K Da | 12/Pk. | N/A | N/A | N/A | ||||||
10203671
![]() |
Sartorius
VS2072 |
0.2 μm | 48/Pk. | 0.2 μm |
825.00 €
Pack of 48 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
10385121
![]() |
Sartorius
VS2022 |
30 kDa | 48/Pk. |
825.00 €
Pack of 48 |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||||
10099490
![]() |
Sartorius
VS2032 |
50 kDa | 48/Pk. |
825.00 €
Pack of 48 |
In Stock
Log in to see stock. ![]() |
|||||||
10672422
![]() |
Sartorius
VS2041 |
100 kDa | 12/Pk. |
230.00 €
Pack of 12 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||||
16656762
|
Sartorius
VS2002 |
10 kDa | 48/Pk. | N/A | N/A | N/A | ||||||
10738231
![]() |
Sartorius
VS2002 |
10 kDa | 48/Pk. |
882.00 €
Pack of 48 |
In Stock
Log in to see stock. ![]() |
|||||||
10039460
|
Sartorius
VS2061 |
1000 kDa | 12/Pk. |
246.00 €
Pack of 12 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||||
10315271
![]() |
Sartorius
VS2012 |
5 kDa | 48/Pk. |
825.00 €
Pack of 48 |
In Stock
Log in to see stock. ![]() |
|||||||
10096881
![]() |
Sartorius
VS2052 |
300 kDa | 48/Pk. |
825.00 €
Pack of 48 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||||
10273521
![]() |
Sartorius
VS2031 |
50 kDa | 12/Pk. |
236.00 €
Pack of 12 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||||
10670333
![]() |
Sartorius
VS2001 |
10 kDa | 12/Pk. |
236.00 €
Pack of 12 |
In Stock
Log in to see stock. ![]() |
|||||||
16636762
|
Sartorius
VS2001 |
10 kDa | 12/Pk. |
247.00 €
Pack of 12 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||||
10012561
![]() |
Sartorius
VS2091 |
3 kDa | 12/Pk. |
236.00 €
Pack of 12 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||||
10325271
![]() |
Sartorius
VS2011 |
5 kDa | 12/Pk. |
236.00 €
Pack of 12 |
In Stock
Log in to see stock. ![]() |
|||||||
10411052
![]() |
Sartorius
VS2051 |
300 kDa | 12/Pk. |
236.00 €
Pack of 12 |
In Stock
Log in to see stock. ![]() |
|||||||
10652232
![]() |
Sartorius
VS2071 |
0.2 μm | 12/Pk. | 0.2 μm | N/A | N/A | N/A | |||||
10638842
![]() |
Sartorius
VS2092 |
3 kDa | 48/Pk. |
825.00 €
Pack of 48 |
In Stock
Log in to see stock. ![]() |
|||||||
Description
Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.
Vivaspin™ 20 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin™ 20 handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Vivaspin™ 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows to monitor the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate. For higher process flexibility, Vivaspin™ 20 is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer is placed on the concentrator and spun to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure. When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, Vivaspin™ 20 can be filled with up to 15ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. Pressure-fugation is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.
- High flux PES membrane
- High retentate recovery > 90 %
- Vertical twin membrane for ultra fast concentration
- Universal rotor compatibility
- Safe sample concentration due to dead-stop volume
- Convenient concentrate recovery by pipetting
- Volume graduation for easy reading/monitoring
- 100-fold concentration with twin vertical membranes
- Additional option for pressurized operation
Rotor Compatibility
Please note: Vivaspin™ 20 (30mm x 116mm) is designed to fit into rotors that can accommodate Falcon 50 ml conical bottom tubes, e.g. Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors; Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors; Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes. These devices are not designed to fit into rotors that only accept round bottom 29mm x 105mm tubes, e.g. Sorvall SS34 or Beckmann JA 20. If your rotor accepts only 29mm x 105mm round bottom tubes, please use the Vivaspin™ 15, which can be used in either round bottom or conical centrifuge tubes
Specifications
Single-use | |
PES | |
Non-sterile | |
116 mm |
Ultrafiltration | |
20 mL | |
14 mL |