Learn More
Cytiva Vivaspin™ 6 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 6 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
Brand: Cytiva VS0601
233.93 EUR valid until 2024-12-31
Use promo code "21930" to get your promotional price.
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 2.5cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
2.5 cm2 | |
6 mL | |
17 mm |
10 kDa | |
25/Pk. | |
122 mm |