missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Vivaspin™ 6 Sample Concentrators

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 6 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

Brand:  Cytiva 28-9322-93

 View more versions of this product

Product Code. 10650561

  • 285.00 € / Pack of 25
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 2.5cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PES
2.5 cm2
6 mL
17 mm
3 kDa
25/Pk.
122 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers