Learn More
Tocris Bioscience™ VL 285
VHL inhibitor
Brand:  Tocris Bioscience™ 7903/10
Product Code. 18754444
  • 186.00 € / 10mg
Explore more special offers
Description
Description
VL 285 is an E3 ubiquitin ligase VHL inhibitor (IC50= 340 nM). VL 285 treatment competitively reduces or reverses target protein degradation by AU-15330 and HaloPROTAC3 Degraders.
Specifications
Specifications
(2S,4R)-4-Hydroxy-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
10 mg
C29H32N4O4S
1448188-57-5
Von Hippel Lindau
0.97