missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ 12-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ 12-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
656.00 € - 713.00 €
Specifications
For Use With | 12 Channel Pipette |
---|---|
Number of Channels | 12 |
Product Line | Acura™ |
Type | Micropipette Volumetric Module |
For Use With (Application) | 12 Channel Pipette |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10749264
|
Socorex™
800.12.010 |
10 μL |
713.00 €
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
10122142
|
Socorex™
800.12.350 |
350 μL |
656.00 €
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
10589482
|
Socorex™
800.12.050 |
50 μL |
656.00 €
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
12 Channel Pipette | |
Acura™ | |
12 Channel Pipette | |
1 Each |
12 | |
Micropipette Volumetric Module | |
Acura™ Electro 926 Micropipette |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title