Learn More
Socorex™ Acura™ 8-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ 8-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
Brand:  Socorex™ 800.08.010
 View more versions of this product
Product Code. 10315732
  • 595.00 € / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
8 Channel Pipette
Acura™
10 μL
Acura™ Electro 926 Micropipette
8
Micropipette Volumetric Module
8 Channel Pipette
1 Each