Learn More
IKA™ VSS 1 Vacuum Safety Set
Condensate trap connected ahead of the vacuum pup to prevent vapors from being released into the air.
Brand:  IKA™ 0020005194
Product Code. 15766678
  • 1053.00 € / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Condensate trap system.
  • Prevents vapors from being released into the air.
  • Suitable for use with RV rotary evaporators and the MVP 10 basic pump:
    • RV 3 V
    • RV 3 V-C
    • RV 3 Flex
    • RV 8 V
    • RV 8 V-C
    • RV 8 Flex
    • RV 10 Auto V
    • RV 10 Auto V-C
    • RV 10 Auto Flex
Specifications
Specifications
For use with RV rotary evaporators and the MVP 10 basic pump