missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VU 0365114, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective positive allosteric modulator of M5 (EC50 values are 2.7 μM for human M5, and >30 μM for M1, M2, M3 and M4 receptors).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 397.35 |
Percent Purity | >98% |
Chemical Name or Material | VU 0365114 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction