Learn More
Emma Safety Footwear Vulcanus XD Safety Shoes
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
Brand:  Emma Safety Footwear MM54086840
 View more versions of this product
Product Code. 15958711
  • 143.00 € / 1 pair
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Velcro over-foot flap
  • Sanitized
Specifications
Specifications
Vulcanus XD
Leather
40