missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining

Easily distinguish cells undergoing apoptosis with Vybrant Apoptosis Assay Kits, which offer dyes, including YO-PRO-1 and Hoechst 33342, for apoptotic and necrotic cell staining.

Brand:  Invitrogen™ V23200

583.30 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 10676523

  • 634.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Apoptotic cells undergo several physiological processes, including changes in cell membrane permeability and chromatin condensation, which can be distinguished using different dyes in apoptotic and necrotic cell staining protocols. Vybrant Apoptosis Assay kits offer streptavidin biotin-annexin V, propidium iodide (PI), YO-PRO-1, and Hoechst 33342 dyes to differentiate live from apoptotic or necrotic cells, which can then be visualized using flow cytometry.

Vybrant Apoptosis Assay kits offer quick and convenient cell staining dye-based methodologies for differentiating apoptotic and necrotic versus healthy cells, which can then be visualized using flow cytometry.

The Vybrant Apoptosis Assay Kit #4 detects apoptosis via changes that occur in cell membrane permeability. This kit contains ready-to-use solutions of both YO-PRO-1 and propidium iodide (PI) nucleic acid stains. YO-PRO-1 stain selectively passes through the plasma membranes of apoptotic cells and labels them with moderate green fluorescence. Necrotic cells are stained fluorescent red with PI.

The Vybrant Apoptosis Assay Kit #5, or the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry, provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. This kit contains ready-to-use solutions of blue-fluorescent Hoechst 33342 dye, which stains the condensed chromatin of apoptotic cells more brightly than the chromatin of non-apoptotic cells, and red-fluorescent PI dye, which stains dead cells.

The Vybrant Apoptosis Assay Kit #6 provides a rapid and convenient assay for apoptosis, using the bright, blue-fluorescent Alexa Fluor 350 conjugate of streptavidin in conjunction with biotin-X annexin V to detect apoptotic cell populations either by flow cytometry or imaging. In addition, the kit includes a ready-to-use solution of the fluorescent red PI nucleic acid–binding dye. PI is impermeant to live cells and apoptotic cells, but stains dead cells with red fluorescence, binding tightly to their nucleic acid. The kit contains an ample amount of each reagent for about 50 flow cytometric assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vybrant Apoptosis Assay Kit #6
Tube
Apoptosis Kit
Alexa Fluor™ 346/442: 346
PI: 535/617
Wet Ice
Fluorescence
Alexa Fluor™ 350 Streptavidin, propidium iodide, Biotin-X Annexin V
Store in refrigerator (2–8°C) and protect from light
50 assays
1 vial of biotin-x annexin V, 1 vial of Alexa Fluor™ 350 streptavidin, 1 vial of propidium iodide, and 1 bottle of 5x annexin binding buffer
UV, 488
Blue, Red
Alexa Fluor™
Flow Cytometer
Alexa Fluor™ Dyes, Biotin & Other Haptens, Other Label(s) or Dye(s)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.