missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining

Product Code. 10709623
Click to view available options
Label or Dye:
Alexa Fluor 350 Streptavidin, Propidium Iodide, Biotin-X Annexin V
Hoechst 33342, Propidium Iodide
YO-PRO-1, propidium iodide
Description:
Vybrant Apoptosis Assay Kit #4
Vybrant Apoptosis Assay Kit #5
Vybrant Apoptosis Assay Kit #6
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 10709623

missing translation for 'mfr': Invitrogen™ V13243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easily distinguish cells undergoing apoptosis with Vybrant Apoptosis Assay Kits, which offer dyes, including YO-PRO-1 and Hoechst 33342, for apoptotic and necrotic cell staining.

Apoptotic cells undergo several physiological processes, including changes in cell membrane permeability and chromatin condensation, which can be distinguished using different dyes in apoptotic and necrotic cell staining protocols. Vybrant Apoptosis Assay kits offer streptavidin biotin-annexin V, propidium iodide (PI), YO-PRO-1, and Hoechst 33342 dyes to differentiate live from apoptotic or necrotic cells, which can then be visualized using flow cytometry.

Vybrant Apoptosis Assay kits offer quick and convenient cell staining dye-based methodologies for differentiating apoptotic and necrotic versus healthy cells, which can then be visualized using flow cytometry.

The Vybrant Apoptosis Assay Kit #4 detects apoptosis via changes that occur in cell membrane permeability. This kit contains ready-to-use solutions of both YO-PRO-1 and propidium iodide (PI) nucleic acid stains. YO-PRO-1 stain selectively passes through the plasma membranes of apoptotic cells and labels them with moderate green fluorescence. Necrotic cells are stained fluorescent red with PI.

The Vybrant Apoptosis Assay Kit #5, or the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry, provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. This kit contains ready-to-use solutions of blue-fluorescent Hoechst 33342 dye, which stains the condensed chromatin of apoptotic cells more brightly than the chromatin of non-apoptotic cells, and red-fluorescent PI dye, which stains dead cells.

The Vybrant Apoptosis Assay Kit #6 provides a rapid and convenient assay for apoptosis, using the bright, blue-fluorescent Alexa Fluor 350 conjugate of streptavidin in conjunction with biotin-X annexin V to detect apoptotic cell populations either by flow cytometry or imaging. In addition, the kit includes a ready-to-use solution of the fluorescent red PI nucleic acid–binding dye. PI is impermeant to live cells and apoptotic cells, but stains dead cells with red fluorescence, binding tightly to their nucleic acid. The kit contains an ample amount of each reagent for about 50 flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Description Vybrant Apoptosis Assay Kit #4
Quantity 200 assays
Format Tube
Kit Contents Contains 1 vial of YO-PRO-1, and 1 vial of propidium iodide.
Product Type Apoptosis Assay Kit
Flow Cytometer Laser Lines 488
Excitation/Emission YO-PRO-1: 491/509
propidium iodide: 535/617
Color Green, Red
Shipping Condition Room Temperature
Product Line Vybrant
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Label or Dye YO-PRO-1, propidium iodide
Label Type Other Label(s) or Dye(s)
Storage Requirements Store in refrigerator (2–8°C) and protect from light
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.