missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Vybrant™ DyeCycle™ Violet/SYTOX™ AADvanced™ Apoptosis Kit, for flow cytometry

Vybrant™ DyeCycle™ Violet/SYTOX™ AADvanced™ Apoptosis Kit, for flow cytometry

Brand:  Molecular Probes™ A35135

Product Code. 10668185

  • 560.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Vybrant™ DyeCycle™ Violet/SYTOX™ AADvanced™ apoptosis kit is used for the identification of apoptotic cells. This kit provides a rapid and convenient assay based upon fluorescence analysis of the compacted state of the nuclear chromatin in apoptotic cells. The kit contains solutions of the cell-permeant Vybrant™ DyeCycle™ Violet stain (Ex/Em max ∼370/440nm) which is excited with UV or violet laser and SYTOX™ AADvanced™ (Ex/Em max ∼546/650 nm) which is excited with a 488 nm laser. SYTOX™ AADvanced™ is used to detect changes in permeability of the plasma membrane therefore allowing identification of dead cells. The staining pattern resulting from simultaneous use of both dyes in combination make it possible to distinguish normal, apoptotic, and dead cell populations by flow cytometry.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 kit
Apoptosis Kit
405, 488
200
Vybrant™
Flow Cytometry
Vybrant DyeCycle Violet, SYTOX AADvanced Dead Cell Stain
Tube
Contains 1 vial of Vybrant™ DyeCycle™ Violet (50 μL, 5mM solution in water); 1 vial SYTOX™ AADvanced™ stain (100 μg); and 1 vial of DMSO (200 μL).

Storage 2-6°C
SYTOX™ AADvanced™: 546⁄647, Vybrant™ DyeCycle™ Violet: 369⁄437
Room Temperature
Fluorescence
Flow Cytometer
Other Label(s) or Dye(s)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.