missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry

Detect apoptosis via flow cytometry with Vybrant FLICA Caspase Assays that detect poly and caspase-3/7/8 activities.

Brand:  Invitrogen™ V35118

340.90 EUR valid until 2025-03-16
Use promo code "24070" to get your promotional price.



 View more versions of this product

Product Code. 10003352

  • 487.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Quickly detect caspase-mediated apoptosis by flow cytometry with Vybrant FAM caspase assay kits for poly-caspases and caspases 3 and 7. The Vybrant FAM caspase assay kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity. Specifically, these caspase assay kits contain engineered FLICA substrates FAM-VAD-FMK (for poly-caspases), FAM-DEVD-FMK (for caspase-3/7), and FAM-LETD-FMK (for caspase-8). They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by flow cytometry.

The Vybrant FAM caspase assay kits for flow cytometry employ a novel approach to detecting active caspases: the kits take advantage of FLICA methodology, which is an affinity label. This affinity label associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase-3/7, and leucine-glutamic acid-threonine-aspartic acid (LETD) for caspase-8. A carboxyfluorescein (FAM) group is attached as a reporter.

The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488 nm/⁄530 nm, 535⁄ nm/6617 nm, and 350 nm/⁄461 nm, respectively. The detection reagents included within the individual Vybrant FAM Poly Caspases Assay, Caspase-3 and -7 Assay, and Caspase-8 Assay apoptosis kits are 488⁄⁄530 FAM-VAD-FMK reagent, 488/⁄530 FAM-DEVD-FMK, and 488/⁄530 FAM-LETD-FMK, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vybrant FAM Caspase-3 and -7 Assay Kit
Tube
Caspase Assay Kit
FAM-DEVD-FMD: 388/530
PI: 535/617
Hoechst 33342: 350/461
Vybrant
Flow Cytometer
Other Label(s) or Dye(s)
25 assays
1 vial FAM-DEVD-FMK caspase-3 and -7 reagent (FLICA reagent, lyophilized solid), 1 vial of Hoechst 33342 (400 μL, 1 mM in water), 1 vial of PI (1 mL, 250 μg/mL in water), 1 vial of DMSO (0.5 mL), 1 bottle of apoptosis fixative solution (10% formaldehyde with methanol in PBS, 6 mL), and 1 bottle of 10x apoptosis wash buffer.
UV, 488
Room Temperature
Fluorescence
FAM, Hoechst 33342, propidium iodide
Store in refrigerator (2–8°C) and protect from light.
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry > 25 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.