missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BICASA Wall Bench MECC Series with Sink
Wall Bench MECC Series with Sink, complete with lateral pedestals, MECC service spine, C-Frame side support, cross-bar, PP sink countertop with 1 basin, water mixer, eyewash and pegboard included.
Brand: BICASA MS.1200.900.930.PP-A
This item is not returnable.
View return policy
Description
- Laboratory benches MECC Series
- PEDESTALS: Aluminum drawn profile (150 x 25 mm and 150 x 50 mm) with vertical slots
- SERVICE SPINE: Modular steel frame assembled to the pedestals at different heights with utility panels horizontal rows
- Trunking structure with passageway for piping and wirings
- UTILITY STRIP: predrilled metal band at countertop height
- UTILITY PANELS: Moulded Polypropylene PP resin cover (V0 flame retarded - IEC 60695-11-10 / -20)
- C-FRAME WORKSTATIONS: certified according to EN 13150:2001 and BRG 234/09.06
- C-shaped steel tubular frame levelling supports - Horizontal cross-bars - Lateral metal sheeting
- COUNTERTOPS: Independent units - installed and fixed above the C-FRAME
- COLORS: METAL: Light Grey matt bonderized Polyurethane powder coating
- UTILITY MECC PANELS: Silver Grey (frontal plate) - ABS Electric panel rear box: Black
- FIRE-RETARDANT PARTICLEBOARD 19 mm panels: Light Grey (2 decorative)
- TAPS: On turret - Epoxy powder coating - Light Grey color
- Locking on utility strip or on countertop with threaded inlet feed sleeve and lock nut
- Fluid identification (handle according to EN13792:2000)
- REAGENT HOLDER SHELF: coated metal - cantilever
- PEGBOARD: Plastic (acrylic stand) panel
Specifications
Wall Bench | |
Anthracite | |
1200 x 900 x 930 mm | |
Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction