Learn More
Vilber Lourmat™ UV Lamp Hand Clamp
Carry your UV lamp easily and conveniently with this specially designed clamp and handle assembly. Vilber Lourmat™ UV Lamp Hand Clamp fits onto 4/6W hand lamps.
Brand:  Vilber Lourmat™ 913020021
Product Code. 11860240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Lamp handle
  • Clip type
  • For UV lamps BVL-4, BVL-8, BVL-6
Specifications
Specifications
Lamp Handle for UV Lamps
1 Ea.