missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Wall Mountable Extraction Unit, without Air Monitoring

For use with tall cabinets, without exhaust/extraction air monitoring.

Brand:  Asecos™ 17178

Product Code. 12658513

  • 801.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Avoids complex wall breakthroughs and expensive extraction piping
  • For use with a range of Asecos cabinets
  • Without extraction air monitoring – the green LED turns off in case of power breakdown
  • Includes installation consoles, flexible hose, hose clips and installation material
Specifications

Specifications

Storage Cabinets
Installation Materials
Extraction Unit
Ventilation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.