missing translation for 'onlineSavingsMsg'
Learn More

Wang resin, 1% cross-linked, 0.8-1.1mmol/g, 200-400 mesh, Thermo Scientific™

Chemical Identifiers

CAS 201058-08-4
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00131778
InChI Key NERFNHBZJXXFGY-UHFFFAOYSA-N
Synonym 4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
PubChem CID 2734007
SMILES CC1=CC=C(COC2=CC=C(CO)C=C2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363926
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L17028.06
5 g N/A
N/A
11373926
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L17028.14
25 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

201058-08-4
228.29
NERFNHBZJXXFGY-UHFFFAOYSA-N
2734007
C15H16O2
MFCD00131778
4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
CC1=CC=C(COC2=CC=C(CO)C=C2)C=C1

Specifications

201058-08-4
C15H16O2
4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
CC1=CC=C(COC2=CC=C(CO)C=C2)C=C1
228.29
Wang resin, 1% cross-linked
200-400 Mesh
MFCD00131778
NERFNHBZJXXFGY-UHFFFAOYSA-N
[4-[(4-methylphenyl)methoxy]phenyl]methanol
2734007
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications