Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
200.00 € - 209.00 €
Specifications
Material | Stainless Steel |
---|---|
Type | Furnace Tongs |
For Use With (Application) | For handling a wide variety of crucibles and to protect the hand against radiation from the furnace |
Quantity | 1 Ea. |
Description
- Manufactured from stainless steel
- For handling a wide variety of crucibles, especially in muffle furnaces
- Operated by thumb action with one hand
Specifications
Stainless Steel | |
Furnace Tongs | |
For handling a wide variety of crucibles and to protect the hand against radiation from the furnace | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
R&L Enterprises™ Ward Pattern Furnace Tongs