missing translation for 'onlineSavingsMsg'
Learn More

Waring™ Stainless Steel Mixer Adapter

For use with 1L containers on 2L base

Brand:  Waring Laboratory Science 21/AD1

Product Code. 12899395

  • 1145.00 € / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.
Specifications

Specifications

Mixer Adapter
1 L and mini containers 4 L Waring and MPR-130-015U
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers