missing translation for 'onlineSavingsMsg'
Learn More

Waring™ Technologi Adapter

For use with 1L and smaller containers on 2L motor base

Brand:  Waring™ 21/AD2

Product Code. 12889395

  • 1145.00 € / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.
Specifications

Specifications

Adapter
For Use of 1L and Smaller Containers on 2L Motor Base
With Glass or Stainless Steel Containers
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers