Learn More
Brady™ Bradylink™ Warning Chains
Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
Brand:  Brady™ 278271
 View more versions of this product
Product Code. 11781076
  • 149.00 € / 1 pack
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
  • Chains are color coded to match Bradylink™ Warning Posts and have a special link design that prevents kinking.
  • Note: Bradylink Chains are not intended to support heavy objects and are recommended for indoor use only.
  • Package includes chain only (stanchions sold separately).
  • The Bradylink™ Warning Post System is non-conductive so it can be safely used in electrically hazardous areas.
Specifications
Specifications
Warning Chain
Yellow
Brady Warning Chain
Traffic Control
50 mm
25 m
1.1 in.
BRADYLINK WARNING CHAIN YELLOW 25Mx8MM
Polyethylene
25 m x 28 mm x 50 mm
1.97 in.
82 ft.
1 Ea.
28 mm