missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wash Buffer, 1.0% Triton™ X-100, pH 8.0

For GST-fusion protein purification

28.00 € - 48.00 €

Specifications

Color Colorless
pH 8
Content And Storage Ambient temperatures
Form Liquid
For Use With (Application) For GST-fusion protein purification
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453199
View Documents
Thermo Scientific™
J60428.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15463199
View Documents
Thermo Scientific™
J60428.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Colorless
Ambient temperatures
For GST-fusion protein purification
8
Liquid
Buffer
Videos
SDS
Documents

Documents

Product Certifications