missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer B for Dynabeads™ mRNA Purification Kits

Product Code. 15677708
Click to view available options
Quantity:
1 x 60 mL
Unit Size:
60mL
This item is not returnable. View return policy

Product Code. 15677708

Brand: Invitrogen™ 11900D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Wash Buffer B is included in the following Dynabeads™ mRNA purification kits.

This Wash Buffer B is included in the following Dynabeads™ mRNA purification kits: Cat. Nos. 61006, 61011, 61012, and 61021. It is made available separately for applications that require more Wash Buffer B than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 60 mL, store at 2°C to 8°C
Packaging Type Kit
Product Type Wash Buffer B
Product Line Dynabeads
Quantity 1 x 60 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.