missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Wash Buffer ENTO-EF

Wash Buffer ENTO-EF 1000 mL to be used with NucleoBond.

Brand:  Macherey-Nagel™ 740381.1000

Product Code. 18091081

  • 114.00 € / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Accessories
Specifications

Specifications

1000 mL
Vaccine-grade plasmid DNA isolation
Wash Buffer
Buffer
1000 mL
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Wash Buffer ENTO-EF > Product Type: Wash Buffer, Quantity: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.